Cesium fluoride ≥99.9%

Supplier: BeanTown Chemical

122170-100G 122170-25G 122170-5G
BT122170-100GEA 169 USD
BT122170-100G BT122170-25G BT122170-5G
Cesium fluoride ≥99.9%
Cesium fluoride

CAS: 13400-13-0; EC No: 236-487-3; MDL No: MFCD00010960; RTECS: FK9650000 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III Powder/Lump; Molecular Formula: CsF; MW: 151.90 Melting Point: 682° Density (g/mL): 4.115 Hygroscopic


  • RTECS: FK9650000
  • Hygroscopic
  • Powder/Lump

Formula: CsF
MW: 151.9
Boiling Pt: 1251
Melting Pt: 680
Density: 4.115 g/mL
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8 III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR