Fullerene C60 ≥99.5%, powder
Supplier: BeanTown Chemical
122150-100MG
122150-1G
122150-5G
BT122150-100MGEA
45.69
USD
BT122150-100MG
BT122150-1G
BT122150-5G
Fullerene C60 ≥99.5%, powder
Fullerene C60
CAS: 99685-96-8; MDL No: MFCD00151408; RTECS: LS9252500
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: C; MW: 12.011
- RTECS: LS9252500
- Powder
Formula:
C₆₀ |
MDL Number:
MFCD00151408 CAS Number: 99685-96-8 UN: 1325 ADR: 4.1,III |
|
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...