(S)-(-)-1,2-Epoxypropane ≥99%

Supplier: BeanTown Chemical

Synonyms: (S)-(-)-Methyloxirane, (S)-(-)-Propylene oxide, (S)-(-)-Propene oxide

122130-100G 122130-25G 122130-5G
BT122130-100GEA 792 USD
BT122130-100G BT122130-25G BT122130-5G
(S)-(-)-1,2-Epoxypropane ≥99%
(S)-(-)-1,2-Epoxypropane

CAS: 16088-62-3; EC No: 240-241-0; MDL No: MFCD00064312; RTECS: UJ2651000 UN No: UN1280; Haz Class: 3; Packing Group: I Liquid; Molecular Formula: C3H6O; MW: 58.08 Boiling Point: 33-34°; Flash point: -37°C (-34°F) Density (g/mL): 0.829; Refractive Index: 1.366


  • RTECS: UJ2651000
  • Refractive index: 1.366
  • Liquid

Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 33…34 °C
Density: 0.829 g/mL
Flash Pt: –37 °C (–34 °F)
MDL Number: MFCD00064312
CAS Number: 16088-62-3
EINECS: 240-241-0
UN: 1280
ADR: 3,I

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR