Nickel(II) nitrate hexahydrate ≥99.995%

Supplier: BeanTown Chemical

122125-250G 122125-50G 122125-5G
BT122125-250GEA 314 USD
BT122125-250G BT122125-50G BT122125-5G
Nickel(II) nitrate hexahydrate ≥99.995%
Nickel(II) nitrate hexahydrate

CAS: 13478-00-7; EC No: 236-068-5; MDL No: MFCD00149805; RTECS: QR7300000 UN No: UN2725; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: Ni(NO3)2·6H2O; MW: 290.80 Melting Point: 56°; Boiling Point: 137° Density (g/mL): 2.05 Hygroscopic


  • RTECS: QR7300000
  • Hygroscopic
  • Crystalline

Formula: Ni(NO₃)₂·6H₂O
Boiling Pt: 137 °C
Melting Pt: 56 °C
Density: 2.05 g/mL
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR