Benzo[b]thiophene ≥98%
Supplier: BeanTown Chemical
Synonyms:
Thianaphthene, 1-Benzothiophene
122115-100G
122115-25G
122115-5G
BT122115-100GEA
257
USD
BT122115-100G
BT122115-25G
BT122115-5G
Benzo[b]thiophene ≥98%
Benzo[b]thiophene
CAS: 95-15-8; EC No: 202-395-7; MDL No: MFCD00005864
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C8H6S; MW: 134.20
Melting Point: 30-33°; Boiling Point: 221-222°
Density (g/mL): 1.149
- Solid
Formula:
C₈H₆S MW: 134.2 g/mol Boiling Pt: 221…222 °C Melting Pt: 30…33 °C Density: 1.149 g/mL |
MDL Number:
MFCD00005864 CAS Number: 95-15-8 EINECS: 202-395-7 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...