1-Bromo-3,5-difluorobenzene ≥98%
Supplier: BeanTown Chemical
CAS: 461-96-1; EC No: 416-710-2; MDL No: MFCD00010305
UN No: UN1993 ; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H3BrF2; MW: 192.99
Boiling Point: 140°; Flash point: 44°C (111°F)
Density (g/mL): 1.676; Refractive Index: 1.499
- Refractive index: 1.499
- Liquid
Formula:
C₆H₃BrF₂ Boiling Pt: 140 °C Density: 1.676 g/mL Flash Pt: 44 °C (111 °F) |
MDL Number:
MFCD00010305 CAS Number: 461-96-1 EINECS: 416-710-2 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...