1-Amino-2,2,2-trifluoroethane ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,2,2-Trifluoroethylamine
122035-1G
122035-25G
122035-5G
BT122035-1GEA
52
USD
BT122035-1G
BT122035-25G
BT122035-5G
1-Amino-2,2,2-trifluoroethane ≥98%
1-Amino-2,2,2-trifluoroethane
CAS: 753-90-2; EC No: 212-041-3; MDL No: MFCD00008132; RTECS: KS0175000
UN No: UN2733; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: CF3CH2NH2; Molecular Formula: C2H4F3N; MW: 99.06
Boiling Point: 36-37°; Flash point: -16°C (3°F)
Density (g/mL): 1.262; Refractive Index: 1.301
- RTECS: KS0175000
- Refractive index: 1.301
- Liquid
Formula:
C₂H₄F₃N MW: 99.06 g/mol Boiling Pt: 36…37 °C Density: 1.262 g/mL Flash Pt: –16 °C (3 °F) |
MDL Number:
MFCD00008132 CAS Number: 753-90-2 EINECS: 212-041-3 UN: 2733 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...