Methylhydrazine sulfate ≥98%

Supplier: BeanTown Chemical

122030-25G 122030-5G
BT122030-25GEA 186.86 USD
BT122030-25G BT122030-5G
Methylhydrazine sulfate ≥98%
Methylhydrazine sulfate

CAS: 302-15-8; EC No: 206-115-4; MDL No: MFCD00035423; RTECS: MV7750000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Linear Formula: CH3NHNH2·H2SO4; Molecular Formula: CH8N2O4S; MW: 144.15 Melting Point: 141-142°


  • RTECS: MV7750000
  • Solid

Formula: CH₆N₂
MW: 144.15 g/mol
Melting Pt: 141…142 °C
MDL Number: MFCD00035423
CAS Number: 302-15-8
EINECS: 206-115-4
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR