Propylbenzene ≥98%

Supplier: BeanTown Chemical

121970-100G 121970-500G
BT121970-100GEA 43 USD
BT121970-100G BT121970-500G
Propylbenzene ≥98%
Propylbenzene

CAS: 103-65-1; EC No: 203-132-9; MDL No: MFCD00009377; RTECS: DA8750000 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: C6H5CH2CH2CH3; Molecular Formula: C9H12; MW: 120.2 Melting Point: -99°; Boiling Point: 159°; Flash point: 42°C (107°F) Density (g/mL): 0.862; Refractive Index: 1.491


  • RTECS: DA8750000
  • Refractive index: 1.491
  • Liquid

Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 159 °C
Melting Pt: –99 °C
Density: 0.862 g/mL
Flash Pt: 42 °C (107 °F)
MDL Number: MFCD00009377
CAS Number: 103-65-1
EINECS: 203-132-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR