Benzaldehyde ≥98%

Supplier: BeanTown Chemical

121935-1KG 121935-250G 121935-3KG
BT121935-1KGEA 53.77 USD
BT121935-1KG BT121935-250G BT121935-3KG
Benzaldehyde ≥98%
Benzaldehyde

CAS: 100-52-7; EC No: 202-860-4; MDL No: MFCD00003299; RTECS: CU4375000 UN No: UN1990; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: C7H6O; MW: 106.12 Melting Point: -26°; Boiling Point: 178-179°; Flash point: 62°C (143°F) Density (g/mL): 1.044; Refractive Index: 1.545 Air Sensitive


  • RTECS: CU4375000
  • Refractive index: 1.545
  • Air Sensitive
  • Liquid

Formula: C₆H₅CHO
MW: 106.12 g/mol
Boiling Pt: 178…179 °C
Melting Pt: –26 °C
Density: 1.044 g/mL
Flash Pt: 62 °C (143 °F)
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98811dd8cad9340872b5c4b5bfb305860eb1f81a&vid=934c1b87225e77b213e66c334d8f27ef4d924847&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR