Gold tribromide ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Gold (III) bromide, Gold(III) bromide

121905-1G 121905-5G
BT121905-1GEA 151.24 USD
BT121905-1G BT121905-5G
Gold tribromide ≥99.99% (trace metals basis)
Gold tribromide

CAS: 10294-28-7; EC No: 233-654-2; MDL No: MFCD00014171 UN No: UN3260; Haz Class: 8; Packing Group: III Granular; Molecular Formula: AuBr3; MW: 436.69


  • Granular

Formula: AuBr₃
MW: 436.68 g/mol
MDL Number: MFCD00014171
CAS Number: 10294-28-7
EINECS: 233-654-2
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR