2,4,6-Trifluorobenzoyl chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,4,6-trifluorobenzoylchloride
121840-1G
121840-5G
BT121840-1GEA
72
USD
BT121840-1G
BT121840-5G
2,4,6-Trifluorobenzoyl chloride ≥97%
2,4,6-Trifluorobenzoyl chloride
CAS: 79538-29-7; MDL No: MFCD00070810
UN No: UN1760; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C7H2ClF3O; MW: 194.54
Boiling Point: 168°
Density (g/mL): 1.499
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₇H₂ClF₃O MW: 194.54 g/mol Boiling Pt: 168 °C Density: 1.499 g/mL |
MDL Number:
MFCD00070810 CAS Number: 79538-29-7 UN: 1760 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...