Methyl methacrylate ≥99% stabilised

Supplier: BeanTown Chemical

Synonyms: Methacrylic acid methyl ester

121790-100ML 121790-2.5L 121790-500ML
BT121790-100MLEA 28.11 USD
BT121790-100ML BT121790-2.5L BT121790-500ML
Methyl methacrylate ≥99% stabilised
Methyl methacrylate

CAS: 80-62-6; EC No: 201-297-1; MDL No: MFCD00008587; RTECS: OZ5075000 UN No: UN1247; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH2=C(CH3)COOCH3; Molecular Formula: C5H8O2; MW: 100.12 Melting Point: -48°; Boiling Point: 100°; Flash point: 9°C (48°F) Density (g/mL): 0.936; Refractive Index: 1.414


  • RTECS: OZ5075000
  • Refractive index: 1.414
  • Liquid

Formula: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Boiling Pt: 100
Melting Pt: -48
Density: 0.936 g/mL
Flash Pt: 9 °C (48 °F)
MDL Number: MFCD00008587
CAS Number: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3 II
Merck Index: 12,06005

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR