N-(3-(Trimethoxysilyl)propyl)ethylenediamine ≥96%

Supplier: BeanTown Chemical

Synonyms: 3-(2-Aminoethylamino)propyltrimethoxysilane, N-(2-Aminoethyl)-3-aminopropyltrimethoxysilane, N-[3-(Trimethoxysilyl)propyl]ethylenediamine

121770-100G 121770-25G 121770-500G
BT121770-100GEA 39 USD
BT121770-100G BT121770-25G BT121770-500G
N-(3-(Trimethoxysilyl)propyl)ethylenediamine ≥96%
N-(3-(Trimethoxysilyl)propyl)ethylenediamine

CAS: 1760-24-3; EC No: 217-164-6; MDL No: MFCD00008173; RTECS: KV7400000 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C8H22N2O3Si; MW: 222.36 Boiling Point: 146°/15 mmHg; Flash point: 68°C (154°F) Density (g/mL): 1.028; Refractive Index: 1.444 Air Sensitive, Moisture Sensitive


  • RTECS: KV7400000
  • Refractive index: 1.444
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
MW: 222.36 g/mol
Boiling Pt: 146 °C/15 mmHg
Melting Pt: 0
Density: 1.028 g/mL
Flash Pt: 68 °C (154 °F)
MDL Number: MFCD00008173
CAS Number: 1760-24-3
EINECS: 217-164-6
ADR: 8 III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR