4-Bromophenol ≥99%

Supplier: BeanTown Chemical

121705-100G 121705-2.5KG 121705-500G
BT121705-100GEA 66.19 USD
BT121705-100G BT121705-2.5KG BT121705-500G
4-Bromophenol ≥99%
4-Bromophenol

CAS: 106-41-2; EC No: 203-394-4; MDL No: MFCD00002313; RTECS: SJ7960000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C6H5BrO; MW: 173.01 Melting Point: 61-64°; Boiling Point: 235-236°


  • RTECS: SJ7960000
  • Solid

Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 235…236 °C
Melting Pt: 61…64 °C
MDL Number: MFCD00002313
CAS Number: 106-41-2
EINECS: 203-394-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR