Fluorene ≥98%
Supplier: BeanTown Chemical
Synonyms:
9H-Fluorene, Diphenylenmethane
121695-100G
121695-25G
121695-500G
BT121695-100GEA
61
USD
BT121695-100G
BT121695-25G
BT121695-500G
Fluorene ≥98%
Fluorene
CAS: 86-73-7; EC No: 201-695-5; MDL No: MFCD00001111; RTECS: LL5670000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Molecular Formula: C13H10; MW: 166.22
Melting Point: 111-114°; Boiling Point: 298°; Flash point: 151°C (304°F)
- RTECS: LL5670000
- Crystalline
Formula:
C₁₃H₁₀ MW: 166.22 g/mol Boiling Pt: 298 °C Melting Pt: 111…114 °C Flash Pt: 151 °C (304 °F) |
MDL Number:
MFCD00001111 CAS Number: 86-73-7 EINECS: 201-695-5 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...