Nickel(II) chloride hexahydrate ≥99.9995%

Supplier: BeanTown Chemical

Synonyms: Nickel dichloride hexahydrate

121645-100G 121645-25G
BT121645-100GEA 452 USD
BT121645-100G BT121645-25G
Nickel(II) chloride hexahydrate ≥99.9995%
Nickel(II) chloride hexahydrate

CAS: 7791-20-0; EC No: 231-743-0; MDL No: MFCD00149809; RTECS: QR6480000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: NiCl2·6H2O; MW: 237.69 Density (g/mL): 1.92 Hygroscopic


  • RTECS: QR6480000
  • Hygroscopic
  • Crystalline

Formula: NiCl₂·6H₂O
Density: 1.92 g/mL
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.9995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR