Zirconium tetrachloride ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Zirconium (IV) chloride
121625-100G
121625-500G
BT121625-100GEA
48.2
USD
BT121625-100G
BT121625-500G
Zirconium tetrachloride ≥99.5%
Zirconium tetrachloride
CAS: 10026-11-6; EC No: 233-058-2; MDL No: MFCD00011306; RTECS: ZH7175000
UN No: UN2503; Haz Class: 8; Packing Group: III
Powder; Linear Formula: ZrCl4; MW: 233.03
Melting Point: 331° (sublimes)
Density (g/mL): 2.803
Moisture Sensitive
- RTECS: ZH7175000
- Moisture Sensitive
- Powder
Formula:
Cl₄Zr MW: 233.03 g/mol Boiling Pt: 3577 Melting Pt: 437 (subl.) Density: 2.803 g/mL |
MDL Number:
MFCD00011306 CAS Number: 10026-11-6 EINECS: 233-058-2 UN: 2503 ADR: 8 III Merck Index: 12,10307 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...