o-Carborane ≥98%
Supplier: BeanTown Chemical
121615-10G
121615-2G
121615-500MG
BT121615-10GEA
749.63
USD
BT121615-10G
BT121615-2G
BT121615-500MG
o-Carborane ≥98%
o-Carborane
CAS: 16872-09-6; EC No: 240-897-8; MDL No: MFCD00012169; RTECS: HS9285000
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: C2H12B10; MW: 144.23
Melting Point: 260°
Density (g/mL): 0.95
- RTECS: HS9285000
- Powder
Formula:
C₂H₁₂B₁₀ MW: 144.23 g/mol Melting Pt: 260 °C Density: 0.95 g/mL |
MDL Number:
MFCD00012169 CAS Number: 16872-09-6 EINECS: 240-897-8 ADR: 4.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...