4-Bromo-o-phenylenediamine ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-Bromo-3,4-diaminobenzene, 4-Bromobenzene-1,2-diamine, 4-Bromo-1,2-phenylenediamine, 4-Bromo-1,2-diaminobenzene, 2-Amino-4-bromophenylamine
121600-100G
121600-25G
121600-5G
BT121600-100GEA
373
USD
BT121600-100G
BT121600-25G
BT121600-5G
4-Bromo-o-phenylenediamine ≥97%
4-Bromo-o-phenylenediamine
CAS: 1575-37-7; MDL No: MFCD02660622
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H7BrN2; MW: 187.04
Melting Point: 65-69°
- Solid
Formula:
C₆H₇BrN₂ Melting Pt: 65…69 °C |
MDL Number:
MFCD02660622 CAS Number: 1575-37-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...