Sodium chloroacetate ≥98%

Supplier: BeanTown Chemical

Synonyms: Chloroacetic acid sodium salt

121550-2.5KG 121550-500G
BT121550-2.5KGEA 69.39 USD
BT121550-2.5KG BT121550-500G
Sodium chloroacetate ≥98%
Sodium chloroacetate

CAS: 3926-62-3; EC No: 223-498-3; MDL No: MFCD00002684; RTECS: AG1400000 UN No: UN2659; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: ClCH2COONa; Molecular Formula: C2H2ClNaO2; MW: 116.48 Melting Point: 199° (decomposes); Flash point: 270°C (518°F)


  • RTECS: AG1400000
  • Powder

Formula: C₂H₂ClNaO₂
MW: 116.48 g/mol
Melting Pt: 199 °C (dec.)
Flash Pt: 270 °C (518 °F)
MDL Number: MFCD00002684
CAS Number: 3926-62-3
EINECS: 223-498-3
UN: 2659
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR