Copper(II) sulfate, anhydrous

Supplier: BeanTown Chemical

Synonyms: Cupric sulphate

121535-100G 121535-2KG 121535-500G
BT121535-100GEA 53.77 USD
BT121535-100G BT121535-2KG BT121535-500G
Copper(II) sulfate, anhydrous
Copper(II) sulphate

CAS: 7758-98-7; EC No: 231-847-6; MDL No: MFCD00010981; RTECS: GL8800000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: CuSO4; MW: 159.60 Melting Point: 560° (decomposes) Density (g/mL): 3.603 Hygroscopic


  • RTECS: GL8800000
  • Hygroscopic
  • Powder

Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 560 °C (dec.)
Density: 3.603 g/mL
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR