3,6-Dichloropyridazine ≥98%

Supplier: BeanTown Chemical

121505-100G 121505-25G 121505-500G
BT121505-100GEA 83 USD
BT121505-100G BT121505-25G BT121505-500G
3,6-Dichloropyridazine ≥98%
3,6-Dichloropyridazine

CAS: 141-30-0; EC No: 205-478-6; MDL No: MFCD00006466; RTECS: UR5886250 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C4H2Cl2N2; MW: 148.98 Melting Point: 65-69°


  • RTECS: UR5886250
  • Solid

Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Melting Pt: 65…69 °C
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR