Barium hydroxide octahydrate ≥98.0% ACS
Supplier: BeanTown Chemical
121485-1KG
121485-250G
BT121485-1KGEA
121
USD
BT121485-1KG
BT121485-250G
Barium hydroxide octahydrate ≥98.0% ACS
Barium hydroxide octahydrate
CAS: 12230-71-6; EC No: 241-234-5; MDL No: MFCD00149152; RTECS: CQ9200000
UN No: UN3262; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: Ba(OH)2·8H2O; MW: 315.46
Melting Point: 78°
Air Sensitive , Hygroscopic
- RTECS: CQ9200000
- Air Sensitive, Hygroscopic
- Crystalline
Formula:
Ba(OH)₂·8H₂O MW: 315.47 g/mol Melting Pt: 78 °C |
MDL Number:
MFCD00149152 CAS Number: 12230-71-6 EINECS: 241-234-5 UN: 3262 ADR: 8,III |
Specification Test Results
Assay | 98.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...