Arsenic(III) oxide 99.95-100.05% ACS primary standard

Supplier: BeanTown Chemical

Synonyms: di-Arsenic trioxide

121465-100G 121465-25G
BT121465-100GEA 359 USD
BT121465-100G BT121465-25G
Arsenic(III) oxide 99.95-100.05% ACS primary standard
Arsenic(III) oxide

CAS: 1327-53-3; EC No: 215-481-4; MDL No: MFCD00003433; RTECS: CG3325000 UN No: UN1561; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: As2O3; MW: 197.84 Boiling Point: 465° (sublimes)


  • RTECS: CG3325000
  • Powder

Formula: As₂O₃
MW: 197.84 g/mol
Boiling Pt: 465 °C (subl.)
MDL Number: MFCD00003433
CAS Number: 1327-53-3
EINECS: 215-481-4
UN: 1561
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99.95-100.05%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR