Disodium sulphide
Supplier: BeanTown Chemical
Synonyms:
Sodium sulphide
121430-100G
121430-25G
121430-5G
BT121430-100GEA
676
USD
BT121430-100G
BT121430-25G
BT121430-5G
Disodium sulphide
Disodium sulphide
CAS: 1313-82-2; EC No: 215-211-5; MDL No: MFCD00003498; RTECS: WE1905000
UN No: UN1385; Haz Class: 4.2; Packing Group: II
Solid; Molecular Formula: Na2S ; MW: 78.04
Melting Point: 950°
Density (g/mL): 1.856
Air Sensitive, Hygroscopic, Light Sensitive
- RTECS: WE1905000
- Air Sensitive, Hygroscopic, Light Sensitive
- Solid
Formula:
Na₂S Melting Pt: 950 °C Density: 1.856 g/mL |
MDL Number:
MFCD00003498 CAS Number: 1313-82-2 EINECS: 215-211-5 UN: 1385 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...