4-(Trifluoromethyl)thiophenol ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol

121375-1G 121375-25G 121375-5G
BT121375-1GEA 72 USD
BT121375-1G BT121375-25G BT121375-5G
4-(Trifluoromethyl)thiophenol ≥97%
4-(Trifluoromethyl)thiophenol

CAS: 825-83-2; MDL No: MFCD00042320 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C7H5F3S; MW: 178.18 Boiling Point: 66-68°/15 mmHg Density (g/mL): 1.3; Refractive Index: 1.492 Air Sensitive


  • Refractive index: 1.492
  • Air Sensitive
  • Liquid

Formula: C₇H₅F₃S
MW: 178.18 g/mol
Boiling Pt: 66…68 °C/15 mmHg
Density: 1.3 g/mL
MDL Number: MFCD00042320
CAS Number: 825-83-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR