2,4,5-Trifluorobenzenesulfonyl chloride ≥97%
Supplier: BeanTown Chemical
121320-1G
121320-5G
BT121320-1GEA
79
USD
BT121320-1G
BT121320-5G
2,4,5-Trifluorobenzenesulfonyl chloride ≥97%
2,4,5-Trifluorobenzenesulphonyl chloride
CAS: 220227-21-4; MDL No: MFCD00077600
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C6H2ClF3O2S; MW: 230.59
Density (g/mL): 1.657
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₆H₂ClF₃O₂S Density: 1.657 g/mL |
MDL Number:
MFCD00077600 CAS Number: 220227-21-4 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...