Triisopropoxyvanadium(V) oxide ≥96%

Supplier: BeanTown Chemical

121315-1G 121315-25G 121315-5G
BT121315-1GEA 23 USD
BT121315-1G BT121315-25G BT121315-5G
Triisopropoxyvanadium(V) oxide ≥96%
Triisopropoxyvanadium(V) oxide

CAS: 5588-84-1; EC No: 226-997-4; MDL No: MFCD00015017 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: V(O)[OCH(CH3)2]3; Molecular Formula: C9H21O4V; MW: 244.20 Melting Point: -12°; Boiling Point: 80-82°/2 mmHg; Flash point: 45°C (113°F) Density (g/mL): 1.035; Refractive Index: 1.479 Moisture Sensitive


  • Refractive index: 1.479
  • Moisture Sensitive
  • Liquid

Formula: C₉H₂₁O₄V
Boiling Pt: 80…82 °C/2 mmHg
Melting Pt: –12 °C
Density: 1.035 g/mL
Flash Pt: 45 °C (113 °F)
MDL Number: MFCD00015017
CAS Number: 5588-84-1
EINECS: 226-997-4
ADR: 3,III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR