Copper(II) sulphate pentahydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

121245-1KG 121245-5KG
BT121245-1KGEA 70.15 USD
BT121245-1KG BT121245-5KG
Copper(II) sulphate pentahydrate ≥99%
Copper(II) sulphate pentahydrate

CAS: 7758-99-8; EC No: 231-847-6; MDL No: MFCD00149681; RTECS: GL8900000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Linear Formula: CuSO4·5H2O; MW: 249.68 Melting Point: 110° (decomposes) Density (g/mL): 2.27


  • RTECS: GL8900000
  • Crystalline

Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Melting Pt: 110 °C (dec.)
Density: 2.27 g/mL
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
Merck Index: 13,02682

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR