1,4-Dibromobenzene ≥98%

Supplier: BeanTown Chemical

121210-100G 121210-250G 121210-50G
BT121210-100GEA 29 USD
BT121210-100G BT121210-250G BT121210-50G
1,4-Dibromobenzene ≥98%
1,4-Dibromobenzene

CAS: 106-37-6; EC No: 203-390-2; MDL No: MFCD00000089; RTECS: CZ1791000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Molecular Formula: C6H4Br2; MW: 235.90 Melting Point: 83-87°; Boiling Point: 219°; Flash point: 100°C (212°F) Density (g/mL): 1.841


  • RTECS: CZ1791000
  • Crystalline

Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 219
Melting Pt: 83…87 °C
Density: 1.841 g/mL
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00000089
CAS Number: 106-37-6
EINECS: 203-390-2
UN: 3077
ADR: 9 III
Merck Index: 13,03042

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR