2-Bromopropane ≥99%

Supplier: BeanTown Chemical

121150-1L 121150-250ML 121150-5L
BT121150-1LEA 139.86 USD
BT121150-1L BT121150-250ML BT121150-5L
2-Bromopropane ≥99%
2-Bromopropane

CAS: 75-26-3; EC No: 200-855-1; MDL No: MFCD00000147; RTECS: TX4111000 UN No: UN2344; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C3H7Br; MW: 122.99 Melting Point: -89°; Boiling Point: 59°; Flash point: 19°C (66°F) Density (g/mL): 1.31; Refractive Index: 1.425


  • RTECS: TX4111000
  • Refractive index: 1.425
  • Liquid

Formula: H₃CBrCHCH₃
MW: 122.99 g/mol
Boiling Pt: 59 °C
Melting Pt: –89 °C
Density: 1.31 g/mL
Flash Pt: 19 °C (66 °F)
MDL Number: MFCD00000147
CAS Number: 75-26-3
EINECS: 200-855-1
UN: 2344
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR