Isoquinoline

Supplier: BeanTown Chemical

121140-100G 121140-2.5KG 121140-500G
BT121140-100GEA 67 USD
BT121140-100G BT121140-2.5KG BT121140-500G
Isoquinoline
Isoquinoline

CAS: 119-65-3; EC No: 204-341-8; MDL No: MFCD00006898; RTECS: NW6825000 UN No: UN2811; Haz Class: 6.1; Packing Group: II Liquid/Low Melting Solid; Molecular Formula: C9H7N ; MW: 129.16 Melting Point: 26-28°; Boiling Point: 242-243°; Flash point: 107°C (225°F) Density (g/mL): 1.099; Refractive Index: 1.623


  • RTECS: NW6825000
  • Refractive index: 1.623
  • Liquid/Low Melting Solid

Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 242…243 °C
Melting Pt: 26…28 °C
Density: 1.099 g/mL
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD00006898
CAS Number: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR