1,1,1-Trifluoro-2-propanol ≥97%
Supplier: BeanTown Chemical
121125-25G
121125-5G
BT121125-25GEA
240
USD
BT121125-25G
BT121125-5G
1,1,1-Trifluoro-2-propanol ≥97%
1,1,1-Trifluoro-2-propanol
CAS: 374-01-6; EC No: 206-773-2; MDL No: MFCD00014403
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CF3CH(OH)CH3; Molecular Formula: C3H5F3O; MW: 114.07
Boiling Point: 81-82°; Flash point: 18°C (64°F)
Density (g/mL): 1.259; Refractive Index: 1.316
- Refractive index: 1.316
- Liquid
Formula:
C₃H₅F₃O MW: 114.07 g/mol Boiling Pt: 81…82 °C Density: 1.259 g/mL Flash Pt: 18 °C (64 °F) |
MDL Number:
MFCD00014403 CAS Number: 374-01-6 EINECS: 206-773-2 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...