Bismuth(III) telluride ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Dibismuth tritelluride
121120-10G
121120-50G
BT121120-10GEA
117
USD
BT121120-10G
BT121120-50G
Bismuth(III) telluride ≥99.999% (trace metals basis)
Bismuth(III) telluride
CAS: 1304-82-1; EC No: 215-135-2; MDL No: MFCD00014201; RTECS: EB3110000
UN No: UN3284; Haz Class: 6.1; Packing Group: III
6mm & Down Pieces ; Molecular Formula: Bi2Te3; MW: 800.76
Melting Point: 585°
Density (g/mL): 7.642
Moisture Sensitive
- RTECS: EB3110000
- Moisture Sensitive
- 6mm & Down Pieces
Formula:
Bi₂Te₃ MW: 800.76 g/mol Melting Pt: 585 °C Density: 7.642 g/mL |
MDL Number:
MFCD00014201 CAS Number: 1304-82-1 EINECS: 215-135-2 UN: 3284 ADR: 6.1,III |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...