Tin(II) chloride hydrate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
|
Danger
|
Synonyms:
Tin dichloride hydrate, Stannous chloride hydrate
121065-100G
121065-25G
121065-5G
BT121065-100GEA
695.4
USD
BT121065-100G
BT121065-25G
BT121065-5G
Tin(II) chloride hydrate ≥99.995% (trace metals basis)
Tin(II) chloride hydrate
CAS: 7772-99-8; EC No: 231-868-0; MDL No: MFCD00011241; RTECS: XP8850000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline/Powder; Linear Formula: SnCl2·xH2O; MW: 189.62 (anhydrous)
Melting Point: 37-38° (decomposes); Boiling Point: 652°
Density (g/mL): 2.71
Air Sensitive, Moisture Sensitive
- RTECS: XP8850000
- Crystalline/Powder
- Air Sensitive, Moisture Sensitive
Formula:
SnCl₂·xH₂O MW: 189.62 g/mol (anhy) Boiling Pt: 652 °C Melting Pt: 37…38 °C (dec.) Density: 2.71 Storage Temperature: Ambient |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 EINECS: 231-868-0 UN: 3260 ADR: 8 III |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...