(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
Supplier: BeanTown Chemical
Synonyms:
(R)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole
121025-1ML
121025-25ML
121025-5ML
BT121025-1MLEA
41
USD
BT121025-1ML
BT121025-25ML
BT121025-5ML
(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine
CAS: 112022-83-0; MDL No: MFCD00078440
UN No: UN1294; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C18H20BNO; MW: 277.17
Flash point: 4°C (39°F)
Density (g/mL): 0.950
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₈H₂₀BNO MW: 277.17 g/mol Density: 0.950 g/mL Flash Pt: 4 °C (39 °F) |
MDL Number:
MFCD00078440 CAS Number: 112022-83-0 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...