Europium ≥99.9% (trace rare earth metals basis), ingot

Supplier: BeanTown Chemical

121010-1G 121010-5G
BT121010-1GEA 147.2 USD
BT121010-1G BT121010-5G
Europium ≥99.9% (trace rare earth metals basis), ingot
Europium

CAS: 7440-53-1; EC No: 231-161-7; MDL No: MFCD00010992 UN No: UN3208; Haz Class: 4.3; Packing Group: II Solid, packaged under mineral oil; Molecular Formula: Eu; MW: 151.96 Melting Point: 822°; Boiling Point: 1527° Density (g/mL): 5.24


  • Solid, packaged under mineral oil

Formula: Eu
MW: 151.96 g/mol
Boiling Pt: 1527 °C
Melting Pt: 822 °C
Density: 5.24 g/mL
MDL Number: MFCD00010992
CAS Number: 7440-53-1
EINECS: 231-161-7
UN: 3208
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR