Glass Fiber Filters, Thick, Advantec MFS
Supplier: Advantec MFS
GD120142MM GB14047MM GA200293MM GD12090MM GB140293MM GD120293MM GB14090MM GA200142MM GD120125MM GD120257MM GB14055MM GB140110MM GB140125MM GA200124MM GA10047MM GD120124MM GB14042.5MM GA200257MM GA100142MM GA100293MM GD12025MM GB14025MM GB140142MM GA20025MM GA20047MM GA20090MM GA10025MM GA10090MM GD12047MM
101102-236PK 193.18 USD
101102-236 101102-342 101102-818 101102-204 101102-424 101102-156 101102-470 101102-530 101102-230 101102-408 101102-556 101102-446 101102-544 101102-684 101102-696 101102-228 101102-514 101102-816 101102-296 101102-564 101102-562 102831-060 102831-062 102831-082 102831-084 102831-086 102831-088 102831-090 102831-092
Glass Fiber Filters, Thick, Advantec MFS
Filters Glass Fiber Filters
Thicker glass fiber filters with high loading capacity and typically faster flow time are recommended for filtering viscous fluids, such as proteinaceous solutions and gels and as prefilter to extend membrane life.
Grade GA100 with fastest filtration speed for precipitated proteins or cells, as well as air pollution monitoring.
Grade GD120 provides high wet strength and very high loading capacity, recommended as prefilter for membrane <1.2 µm.
Grade GB140 has a slower filtration speed, high dust holding capacity, and can be used in suspended solid analysis and industrial waste.
Grade GA200 thick filter with high loading capacity for liquid sugars and gels.
Binder-free.
Información para pedidos: Other diameters available.