Bis(cyclopentadienyl)zirconium hydridochloride ≥96.0%

Supplier: TCI America

Synonyms: Bis(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz's Reagent, Bis(cyclopentadienyl)zirconium chloride hydride

Z0010-1G Z0010-25G Z0010-5G
TCZ0010-1GEA 40 USD
TCZ0010-1G TCZ0010-25G TCZ0010-5G
Bis(cyclopentadienyl)zirconium hydridochloride ≥96.0%
Bis(cyclopentadienyl)zirconium hydridochloride

CAS Number: 37342-97-5
MDL Number: MFCD02089401
Molecular Formula: C10H11ClZr
Molecular Weight: 257.87
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Reduction, Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Bis(cyclopentadienyl)zirconium Chloride Hydride, Schwartz's Reagent

Formula: C₁₀H₁₁ClZr
MW: 257.87 g/mol
MDL Number: MFCD02089401
CAS Number: 37342-97-5
UN: 3131
ADR: 4.3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=409d1938bb6f710df7793330b3f24aba3b8a2467&vid=409d1938bb6f710df7793330b3f24aba3b8a2467&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR