Zirconocene bis(trifluoromethanesulfonate) tetrahydrofuran adduct ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bis(cyclopentadienyl)zirconium Bis(trifluoromethanesulphonate), Zirconocene Bis(triflate)

Z0009-1G Z0009-5G
TCZ0009-1GEA 141.44 USD
TCZ0009-1G TCZ0009-5G
Zirconocene bis(trifluoromethanesulfonate) tetrahydrofuran adduct ≥97.0% (by titrimetric analysis)
Zirconocene bis(trifluoromethanesulfonate) tetrahydrofuran

CAS Number: 89672-77-5
MDL Number: MFCD00192583
Molecular Formula: C16H18F6O7S2Zr
Molecular Weight: 591.65
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Flash Point (°C): 79


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Homogeneous Catalysts, Homogeneous Catalysts
Other Product Names / Synonyms: Bis(cyclopentadienyl)zirconium Bis(trifluoromethanesulfonate), Zirconocene Bis(triflate)

Formula: C₁₆H₁₈F₆O₇S₂Zr
MW: 591.65 g/mol
Flash Pt: 79 °C
MDL Number: MFCD00192583
CAS Number: 89672-77-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec1a7d384ce61b00795c74094c41bd5e3c097fc0&vid=0e595d2e5ee288c6abb71ea3418b7483a125a717&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR