Xylazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine Hydrochloride, 2-[(2,6-Dimethylphenyl)amino]-5,6-dihydro-4H-1,3-thiazine Hydrochloride
CAS Number: 23076-35-9
MDL Number: MFCD00058196
Molecular Formula: C12H16N2S
Molecular Weight: 256.79
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine Hydrochloride, 2-[(2,6-Dimethylphenyl)amino]-5,6-dihydro-4H-1,3-thiazine Hydrochloride
Formula:
C₁₂H₁₆N₂S MW: 256.79 g/mol |
MDL Number:
MFCD00058196 CAS Number: 23076-35-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...