o-Tolylacetonitrile ≥98.0%
Supplier: TCI America
Synonyms:
(2-Methylphenyl)acetonitrile, 2-(o-Tolyl)acetonitrile, 2-Methylbenzyl cyanide
X0045-25ML
X0045-100ML
TCX0045-25MLEA
76
USD
TCX0045-25ML
TCX0045-100ML
o-Tolylacetonitrile ≥98.0%
o-Tolylacetonitrile
CAS Number: 22364-68-7
MDL Number: MFCD00001904
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: 2-Methylbenzyl Cyanide, o-Tolylacetonitrile
Formula:
C₉H₉N Boiling Pt: 120 °C Density: 1.01 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00001904 CAS Number: 22364-68-7 UN: 3276 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...