Warfarin sodium ≥98.0% (by HPLC) (contains Isopropyl Alcohol)
Supplier: TCI America
Synonyms:
3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
W0005-25G
W0005-5G
TCW0005-25GEA
198.32
USD
TCW0005-25G
TCW0005-5G
Warfarin sodium ≥98.0% (by HPLC) (contains Isopropyl Alcohol)
Warfarin sodium
CAS Number: 129-06-6
MDL Number: MFCD00083223
Molecular Formula: C19H16O4
Molecular Weight: 330.31
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Product Categories: Bioscience, Antithrombotic Ingredients, Phenylpropanoids
Other Product Names / Synonyms: 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin Sodiuim Salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin Sodium Salt
Formula:
C19H16NaO4 MW: 331.33 g/mol |
MDL Number:
MFCD00083223 CAS Number: 129-06-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...