3-Vinylpyridine ≥96.0% (by GC, titration analysis) stabilized

Supplier: TCI America

V0099-1G V0099-5G
TCV0099-1GEA 205 USD
TCV0099-1G TCV0099-5G
3-Vinylpyridine ≥96.0% (by GC, titration analysis) stabilized
3-Vinylpyridine

(stabilized with TBC)
CAS Number: 1121-55-7
MDL Number: MFCD02178019
Molecular Formula: C7H7N
Molecular Weight: 105.14
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 82
Specific Gravity (20/20): 0.98
Storage Temperature: <0°C

Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 82 °C
Density: 0.98 (20 °C)
MDL Number: MFCD02178019
CAS Number: 1121-55-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28cbc91cabe3024a84dd9e02cba9eed6cbe5f080&vid=28cbc91cabe3024a84dd9e02cba9eed6cbe5f080&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR