Vinyltrimethylsilane ≥97.0% (by GC)

Supplier: TCI America

V0067-100ML V0067-25ML
TCV0067-100MLEA 278 USD
TCV0067-100ML TCV0067-25ML
Vinyltrimethylsilane ≥97.0% (by GC)
Vinyltrimethylsilane

CAS Number: 754-05-2
MDL Number: MFCD00008606
Molecular Formula: C5H12Si
Molecular Weight: 100.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 56
Flash Point (°C): -24
Specific Gravity (20/20): 0.69


Product Categories: Synthetic Organic Chemistry, Vinylation, Materials Chemistry, Monomers
Other Product Names / Synonyms: Trimethylvinylsilane

Formula: C₅H₁₂Si
Boiling Pt: 56 °C
Density: 0.69 (20 °C)
Flash Pt: –24 °C
MDL Number: MFCD00008606
CAS Number: 754-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=987618ad45b6bd7d71c074ca9ab68d4c7244d7db&vid=987618ad45b6bd7d71c074ca9ab68d4c7244d7db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR