Trichloro(6-phenylhexyl)silane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (6-Phenylhexyl)trichlorosilane

T2854-25G T2854-5G
TCT2854-25GEA 672 USD
TCT2854-25G TCT2854-5G
Trichloro(6-phenylhexyl)silane ≥98.0% (by GC)
Trichloro(6-phenylhexyl)silane

CAS Number: 18035-33-1
MDL Number: MFCD09953885
Molecular Formula: C12H17Cl3Si
Molecular Weight: 295.70
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 92
Specific Gravity (20/20): 1.15


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: (6-Phenylhexyl)trichlorosilane

Formula: C₁₂H₁₇Cl₃Si
MW: 295.70 g/mol
Boiling Pt: 92 °C
Density: 1.15 (20 °C)
MDL Number: MFCD09953885
CAS Number: 18035-33-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=738946b212ba9ae9720bd1ead3c9518fc7e59701&vid=738946b212ba9ae9720bd1ead3c9518fc7e59701&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR