Trimethyl[3-(trimethoxysilyl)propyl]ammonium chloride ∼50% in methanol

Supplier: TCI America

T2796-10G T2796-25G
TCT2796-10GEA 43.76 USD
TCT2796-10G TCT2796-25G
Trimethyl[3-(trimethoxysilyl)propyl]ammonium chloride ∼50% in methanol
Trimethyl[3-(trimethoxysilyl)propyl]ammonium chloride

CAS Number: 35141-36-7
MDL Number: MFCD00054225
Molecular Formula: C9H24ClNO3Si
Molecular Weight: 257.83
Form: Clear Liquid
Color: Colorless

Formula: C₉H₂₄ClNO₃Si
MDL Number: MFCD00054225
CAS Number: 35141-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13ec0820053f5bed15e0ee2bf340942c36e8e56b&vid=18f9529a683467c03b71604be80107751056dc83&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR