3-(Trifluoromethyl)thiophenol ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Mercaptobenzotrifluoride, 3-(Trifluoromethyl)benzenethiol, 3-(Trifluoromethyl)benzene-1-thiol

T2780-25G T2780-5G
TCT2780-25GEA 447 USD
TCT2780-25G TCT2780-5G
3-(Trifluoromethyl)thiophenol ≥95.0% (by GC, titration analysis)
3-(Trifluoromethyl)thiophenol

CAS Number: 937-00-8
MDL Number: MFCD00041142
Molecular Formula: C7H5F3S
Molecular Weight: 178.17
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 163
Specific Gravity (20/20): 1.32
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Trifluoromethyl)thiophenol, 3-Mercaptobenzotrifluoride

Formula: C₇H₅F₃S
MW: 178.18 g/mol
Boiling Pt: 163 °C
Density: 1.32 (20 °C)
MDL Number: MFCD00041142
CAS Number: 937-00-8
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc1e57a9c6a94087cef20cf7d4152f639929e55c&vid=bc1e57a9c6a94087cef20cf7d4152f639929e55c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR