1H-Tetrazole-5-acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T2737-25G
T2737-5G
TCT2737-25GEA
956
USD
TCT2737-25G
TCT2737-5G
1H-Tetrazole-5-acetic acid ≥98.0% (by GC, titration analysis)
1H-Tetrazole-5-acetic acid
CAS Number: 21743-75-9
MDL Number: MFCD03094010
Molecular Formula: C3H4N4O2
Molecular Weight: 128.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 177
Formula:
C₃H₄N₄O₂ MW: 128.09 g/mol Melting Pt: 177 °C |
MDL Number:
MFCD03094010 CAS Number: 21743-75-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...